Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003134.1 | NP_001003134.1 | 12 - 13 | 1618 | 122 | |
XM_005638735.3 | XP_005638792.1 | 13 - 14 | 1706 | 122 | ||
XM_022413059.1 | XP_022268767.1 | 13 - 14 | 1675 | 122 | ||
GenBank mRNA | AF239823.1 | - | 12 - 13 | 1618 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |