Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003352.1 | NP_001003352.1 | 1 - 2 | 938 | 95 | |
XM_005636006.3 | XP_005636063.1 | 1 - 2 | 941 | 95 | ||
XM_005636008.3 | XP_005636065.1 | 1 - 2 | 941 | 95 | ||
XM_005636009.3 | XP_005636066.1 | 1 - 2 | 941 | 95 | ||
XM_005636010.3 | XP_005636067.1 | 1 - 2 | 941 | 95 | ||
XM_014107712.2 | XP_013963187.1 | 1 - 2 | 941 | 95 | ||
GenBank mRNA | AF346625.1 | - | 1 - 2 | 938 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |