Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003061.1 | NP_001003061.1 | 5 - 6 | 841 | 81 | |
XM_005622224.3 | XP_005622281.1 | 6 - 7 | 1041 | 81 | ||
XM_022420726.1 | XP_022276434.1 | 6 - 7 | 861 | 81 | ||
GenBank mRNA | AF384053.1 | - | 5 - 6 | 841 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |