Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633719.3 | XP_005633776.1 | 2 | 1078 | 92 | |
GenBank mRNA | AY929390.1 | - | 1 | 271 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Inventoried Both primers and probe map within a single exon |