Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012619.1 | NP_001012637.1 | 2 - 3 | 588 | 69 | |
GenBank mRNA | AJ884915.1 | - | 2 - 3 | 588 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Canis familiaris Probe spans exons |