Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003223.1 | NP_001003223.1 | 6 - 7 | 664 | 83 | |
XM_014114387.1 | XP_013969862.1 | 6 - 7 | 945 | 83 | ||
XM_022419477.1 | XP_022275185.1 | 6 - 7 | 875 | 83 | ||
GenBank mRNA | AY069922.1 | - | 6 - 7 | 640 | 83 | |
D29807.1 | - | 6 - 7 | 664 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Inventoried Probe spans exons |