Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003357.2 | NP_001003357.2 | 7 - 8 | 681 | 79 | |
NM_001313818.1 | NP_001300747.1 | 6 - 7 | 574 | 79 | ||
GenBank mRNA | AY283372.1 | - | 7 - 8 | 532 | 79 | |
DN394227.1 | - | 7 - 8 | 691 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |