Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629550.3 | XP_005629607.1 | 551 | 83 | ||
XM_005629551.3 | XP_005629608.1 | 719 | 83 | |||
XM_022403735.1 | XP_022259443.1 | 719 | 83 | |||
XM_022403736.1 | XP_022259444.1 | 719 | 83 | |||
GenBank mRNA | AY545218.2 | - | 806 | 83 | ||
AY545219.2 | - | 886 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |