Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003194.2 | NP_001003194.1 | 3 - 4 | 368 | 63 | |
XM_005640752.2 | XP_005640809.1 | 3 - 4 | 446 | 63 | ||
XM_022415287.1 | XP_022270995.1 | 3 - 4 | 464 | 63 | ||
GenBank mRNA | AY630442.1 | - | 3 - 4 | 368 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |