Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012344.1 | NP_001012344.1 | 597 | 60 | ||
XM_005617078.3 | XP_005617135.1 | 920 | 60 | |||
XM_005617079.3 | XP_005617136.1 | 763 | 60 | |||
GenBank mRNA | AY875970.1 | - | 597 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris |