Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003319.2 | NP_001003319.2 | 1 - 2 | 30 | 61 | |
GenBank mRNA | DN319335.1 | - | 1 - 2 | 30 | 61 | |
M35302.1 | - | 1 - 2 | 36 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |