Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003272.1 | NP_001003272.1 | 5 - 6 | 635 | 101 | |
XM_005623180.3 | XP_005623237.1 | 4 - 5 | 835 | 101 | ||
XM_005623181.3 | XP_005623238.1 | 4 - 5 | 441 | 101 | ||
GenBank mRNA | X16318.1 | - | 635 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |