Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252217.1 | NP_001239146.1 | 2 - 3 | 217 | 113 | |
GenBank mRNA | DN393350.1 | - | 2 - 3 | 217 | 113 | |
DN434916.1 | - | 2 - 3 | 202 | 113 | ||
DQ403010.1 | - | 2 - 3 | 174 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |