Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431728.4 | XP_003431776.1 | 1 - 2 | 248 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |