Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433090.4 | XP_003433138.1 | 5 - 6 | 843 | 70 | |
XM_005634127.3 | XP_005634184.1 | 5 - 6 | 843 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |