Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633172.2 | XP_005633229.1 | 6 - 7 | 786 | 114 | |
XM_005633173.2 | XP_005633230.1 | 5 - 6 | 714 | 114 | ||
XM_022407331.1 | XP_022263039.1 | 6 - 7 | 848 | 114 | ||
XM_022407332.1 | XP_022263040.1 | 6 - 7 | 852 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |