Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079767.1 | NP_001073235.1 | 429 | 83 | ||
XM_022410149.1 | XP_022265857.1 | 387 | 83 | |||
GenBank mRNA | EF088670.1 | - | 429 | 83 |