Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636115.3 | XP_005636172.1 | 1074 | 110 | ||
XM_005636116.3 | XP_005636173.1 | 1074 | 110 | |||
XM_022410726.1 | XP_022266434.1 | 1075 | 110 | |||
XM_022410727.1 | XP_022266435.1 | 1073 | 110 | |||
XM_534647.6 | XP_534647.3 | 1071 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |