Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636893.3 | XP_005636950.1 | 1 - 2 | 241 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |