Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022411295.1 | XP_022267003.1 | 7 - 8 | 1513 | 73 | |
XM_022411297.1 | XP_022267005.1 | 7 - 8 | 1513 | 73 | ||
XM_022411300.1 | XP_022267008.1 | 7 - 8 | 1513 | 73 | ||
XM_022411303.1 | XP_022267011.1 | 7 - 8 | 1513 | 73 | ||
XM_535301.6 | XP_535301.4 | 7 - 8 | 1510 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |