Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535723.6 | XP_535723.3 | 1 | 373 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |