Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB001388.1 | - | 1 - 2 | 114 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |