Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624863.3 | XP_005624920.1 | 4 - 5 | 583 | 80 | |
XM_014116785.2 | XP_013972260.1 | 3 - 4 | 527 | 80 | ||
XM_022423346.1 | XP_022279054.1 | 2 - 3 | 357 | 80 | ||
XM_863139.5 | XP_868232.1 | 3 - 4 | 443 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |