Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005633033.3 | XP_005633090.1 | 7 - 8 | 4866 | 63 | |
XM_005633034.3 | XP_005633091.1 | 7 - 8 | 4863 | 63 | ||
XM_005633035.3 | XP_005633092.1 | 8 - 9 | 4668 | 63 | ||
XM_005633036.3 | XP_005633093.1 | 8 - 9 | 4668 | 63 | ||
XM_005633039.3 | XP_005633096.1 | 8 - 9 | 4668 | 63 | ||
XM_005633040.3 | XP_005633097.1 | 8 - 9 | 4668 | 63 | ||
XM_005633041.3 | XP_005633098.1 | 8 - 9 | 4668 | 63 | ||
XM_005633043.3 | XP_005633100.1 | 8 - 9 | 4569 | 63 | ||
XM_005633046.3 | XP_005633103.1 | 8 - 9 | 4470 | 63 | ||
XM_005633048.3 | XP_005633105.1 | 8 - 9 | 4272 | 63 | ||
XM_005633050.3 | XP_005633107.1 | 8 - 9 | 3975 | 63 | ||
XM_005633051.3 | XP_005633108.1 | 8 - 9 | 3975 | 63 | ||
XM_005633052.3 | XP_005633109.1 | 8 - 9 | 3875 | 63 | ||
XM_005633053.3 | XP_005633110.1 | 8 - 9 | 3875 | 63 | ||
XM_005633054.3 | XP_005633111.1 | 8 - 9 | 3876 | 63 | ||
XM_022407198.1 | XP_022262906.1 | 8 - 9 | 4569 | 63 | ||
XM_022407199.1 | XP_022262907.1 | 8 - 9 | 4073 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |