Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639455.4 | XP_003639503.1 | 4 - 5 | 551 | 73 | |
XM_022426948.1 | XP_022282656.1 | 5 - 6 | 1045 | 73 | ||
XM_022426949.1 | XP_022282657.1 | 6 - 7 | 845 | 73 | ||
XM_022426950.1 | XP_022282658.1 | 6 - 7 | 823 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |