Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630093.2 | XP_005630150.1 | 448 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |