Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427573.1 | XP_022283281.1 | 11 - 12 | 1408 | 148 | |
XM_532514.5 | XP_532514.3 | 11 - 12 | 1262 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |