Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313863.1 | NP_001300792.1 | 4 - 5 | 350 | 145 | |
XM_022425755.1 | XP_022281463.1 | 4 - 5 | 424 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |