Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120475.2 | XP_013975950.1 | 663 | 73 | ||
XM_022404403.1 | XP_022260111.1 | 702 | 73 | |||
XM_022404404.1 | XP_022260112.1 | 480 | 73 | |||
XM_022404405.1 | XP_022260113.1 | 490 | 73 | |||
XM_022404406.1 | XP_022260114.1 | 503 | 73 | |||
XM_022404407.1 | XP_022260115.1 | 417 | 73 | |||
XM_022404408.1 | XP_022260116.1 | 380 | 73 | |||
XM_022404409.1 | XP_022260117.1 | 523 | 73 | |||
XM_532856.7 | XP_532856.3 | 786 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |