Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431744.4 | XP_003431792.1 | 4 - 5 | 408 | 61 | |
XM_532172.6 | XP_532172.3 | 5 - 6 | 436 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |