Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624229.3 | XP_005624286.1 | 867 | 62 | ||
XM_005624230.2 | XP_005624287.1 | 858 | 62 | |||
XM_014116390.2 | XP_013971865.1 | 867 | 62 | |||
XM_022422804.1 | XP_022278512.1 | 867 | 62 | |||
XM_022422805.1 | XP_022278513.1 | 867 | 62 | |||
XM_022422806.1 | XP_022278514.1 | 867 | 62 | |||
XM_537604.6 | XP_537604.3 | 867 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |