Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022418903.1 | XP_022274611.1 | 1294 | 85 | ||
XM_536708.6 | XP_536708.2 | 1449 | 85 |