Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620092.3 | XP_005620149.1 | 3 - 4 | 1105 | 93 | |
XM_014113621.2 | XP_013969096.1 | 3 - 4 | 1105 | 93 | ||
XM_022418623.1 | XP_022274331.1 | 4 - 5 | 1120 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |