Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536790.6 | XP_536790.2 | 1 - 2 | 173 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |