Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620181.2 | XP_005620238.1 | 2 - 3 | 501 | 80 | |
XM_005620182.3 | XP_005620239.1 | 3 - 4 | 359 | 80 | ||
XM_005620183.2 | XP_005620240.1 | 3 - 4 | 279 | 80 | ||
XM_536670.5 | XP_536670.2 | 2 - 3 | 275 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |