Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538032.6 | XP_538032.1 | 1 - 2 | 486 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |