Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537012.6 | XP_537012.1 | 1 - 2 | 261 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |