Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641417.3 | XP_005641474.2 | 7 - 8 | 1058 | 64 | |
XM_014111773.2 | XP_013967248.1 | 7 - 8 | 1062 | 64 | ||
XM_014111774.2 | XP_013967249.1 | 7 - 8 | 1055 | 64 | ||
XM_022415754.1 | XP_022271462.1 | 6 - 7 | 935 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |