Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628649.3 | XP_005628706.1 | 5 - 6 | 507 | 98 | |
XM_022427500.1 | XP_022283208.1 | 4 - 5 | 1014 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |