Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGREF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404564.1 | XP_022260272.1 | 1 - 2 | 143 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |