Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615432.3 | XP_005615489.1 | 372 | 85 | ||
XM_005615433.3 | XP_005615490.1 | 380 | 85 | |||
XM_005615434.3 | XP_005615491.1 | 358 | 85 | |||
XM_022417317.1 | XP_022273025.1 | 384 | 85 | |||
XM_022417320.1 | XP_022273028.1 | 376 | 85 | |||
XM_022417323.1 | XP_022273031.1 | 372 | 85 |