Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284434.1 | NP_001271363.1 | 3 - 4 | 213 | 72 | |
XM_005627974.3 | XP_005628031.1 | 4 - 5 | 306 | 72 | ||
GenBank mRNA | JX987297.1 | - | 3 - 4 | 213 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |