Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616898.2 | XP_005616955.2 | 3 - 4 | 245 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |