Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100856603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404359.1 | XP_022260067.1 | 3 - 4 | 1326 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |