Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617684.3 | XP_005617741.1 | 1 - 2 | 184 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |