Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618812.3 | XP_005618869.1 | 314 | 143 | ||
XM_022417409.1 | XP_022273117.1 | 314 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |