Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637824.3 | XP_005637881.1 | 6 - 7 | 1923 | 92 | |
XM_005637826.3 | XP_005637883.1 | 6 - 7 | 2117 | 92 | ||
XM_005637827.3 | XP_005637884.1 | 6 - 7 | 1966 | 92 | ||
XM_022411597.1 | XP_022267305.1 | 7 - 8 | 2244 | 92 | ||
XM_022411598.1 | XP_022267306.1 | 5 - 6 | 1819 | 92 | ||
XM_022411599.1 | XP_022267307.1 | 6 - 7 | 2140 | 92 | ||
XM_544058.7 | XP_544058.4 | 5 - 6 | 2013 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |