Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639508.3 | XP_005639565.2 | 3 - 4 | 2566 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |