Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_543740.6 | XP_543740.1 | 4 - 5 | 1313 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |