Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405900.1 | XP_022261608.1 | 1 | 239 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |